missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Gulonic acid gamma-lactone, 95+%

Product Code. 11916191 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11916191 100 g Glass Bottle 100g
10327811 25 g Glass Bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11916191 Supplier Thermo Scientific Acros Supplier No. 255571000
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1128-23-0 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-23-0
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
InChI Key SXZYCXMUPBBULW-SKNVOMKLSA-N
Synonym l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone
PubChem CID 439373
ChEBI CHEBI:17587
IUPAC Name (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Melting Point 183.0°C to 185.0°C
Density 1.344g/mL
Loss on Drying 0.5% max. (1 g, 105°C, 3 hrs)
Infrared Spectrum Authentic
Assay Percent Range 95% min. (ELSD) (HPLC)
Packaging Glass Bottle
Quantity 100 g
Specific Rotation Condition + 54.50 (57.50°C c=2,water)
Specific Rotation + 54.50
Formula Weight 178.14
Percent Purity 95+%
Physical Form Viscous Solution
Chemical Name or Material L(+)-Gulonic acid γ-lactone
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.