missing translation for 'onlineSavingsMsg'
Learn More

L-Gulonic acid-1,4-lactone, 95%

Product Code. 15438499
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15438499 5 g 5g
15448499 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15438499 Supplier Thermo Scientific Alfa Aesar Supplier No. J66934.06
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1128-23-0 | C6H10O6 | 178.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1128-23-0
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00064331
InChI Key SXZYCXMUPBBULW-SKNVOMKLSA-N
Synonym l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone
PubChem CID 439373
ChEBI CHEBI:17587
IUPAC Name (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Melting Point 184°C to 190°C
Color White
Quantity 5 g
Beilstein 83002
Solubility Information Soluble in water
Optical Rotation +54.5° (c=4 in Water)
Formula Weight 178.14
Percent Purity 95%
Physical Form Powder
Chemical Name or Material L-Gulonic acid-1,4-lactone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.