missing translation for 'onlineSavingsMsg'
Learn More

L-Glutathione reduced, MedChemExpress

Product Code. p-200101075
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16431168

Brand: Medchem Express HYD0187

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-Glutathione reduced (GSH; γ-L-Glutamyl-L-cysteinyl-glycine) is an endogenous antioxidant and is capable of scavenging oxygen-derived free radicals.

L-Glutathione reduced (GSH

Specifications

Chemical Name or Material L-Glutathione reduced
CAS 70-18-8
Color White
Purity Grade Notes Research
Molecular Formula C10H17N3O6S
Quantity 5 g
Synonym GSH γ-L-Glutamyl-L-cysteinyl-glycine
Solubility Information H2O : 62.5 mg/mL (203.37 mM; Need ultrasonic)
SMILES O=C(O)CNC([C@H](CS)NC(CC[C@H](N)C(O)=O)=O)=O
Molecular Weight (g/mol) 307.32
Formula Weight 307.32
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.