missing translation for 'onlineSavingsMsg'
Learn More

L-Glutathione reduced, MedChemExpress

Product Code. 16431168
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431168 5 g 5g
16402419 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16431168 Supplier Medchem Express Supplier No. HYD0187
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Glutathione reduced (GSH; γ-L-Glutamyl-L-cysteinyl-glycine) is an endogenous antioxidant and is capable of scavenging oxygen-derived free radicals.

L-Glutathione reduced (GSH

Specifications

Chemical Name or Material L-Glutathione reduced
CAS 70-18-8
Color White
Purity Grade Notes Research
Molecular Formula C10H17N3O6S
Quantity 5 g
Synonym GSH γ-L-Glutamyl-L-cysteinyl-glycine
Solubility Information H2O : 62.5 mg/mL (203.37 mM; Need ultrasonic)
SMILES O=C(O)CNC([C@H](CS)NC(CC[C@H](N)C(O)=O)=O)=O
Molecular Weight (g/mol) 307.32
Formula Weight 307.32
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.