missing translation for 'onlineSavingsMsg'
Learn More

L(-)-Glutathione, oxidized hydrate, 95%

Product Code. 10697744 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10697744 25 g Glass bottle 25g
10022001 1 g Glass bottle 1g
10683342 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10697744 Supplier Thermo Scientific Acros Supplier No. 343840250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 27025-41-8 | C20H32N6O12S2 | 612.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27025-41-8
Molecular Formula C20H32N6O12S2
Molecular Weight (g/mol) 612.63
MDL Number MFCD00150701
InChI Key YPZRWBKMTBYPTK-IOKZUGQQNA-N
Synonym oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
PubChem CID 65359
ChEBI CHEBI:17858
IUPAC Name (2S)-2-amino-5-[[(2R)-3-[[(2R)-2-[[(4S)-4-amino-4-carboxybutanoyl]amino]-3-(carboxymethylamino)-3-oxopropyl]disulfanyl]-1-(carboxymethylamino)-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
SMILES N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

Specifications

Melting Point 178.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Merck Index 15, 4511
Specific Rotation Condition − 99.00 (20.00°C c=4,water)
Specific Rotation -92° to -106° (20°C, 589nm) (c=2, H2O)
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in most common organic solvents
Formula Weight 612.64
Percent Purity 95%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material L(-)-Glutathione, oxidized hydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.