missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L(-)-Glutathione, oxidized, 98%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 mg
5 g
Packaging:
Glass bottle
Unit Size:
500mg
5g
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 178.0°C |
| Color | White |
| Ignition Residue | 0.5% max. (800°C) |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97.5% min. (HPLC) |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Beilstein | 04, IV, 3168 |
| Merck Index | 15, 4511 |
| Specific Rotation Condition | −99.00 (20.00°C c=4,water) |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction