missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, U.S.P. - F.C.C., J.T. Baker™

Product Code. 12694725
Change view
Click to view available options
Quantity:
1 kg
Unit Size:
1kg
Product Code. Quantity unitSize
12694725 1 kg 1kg
1 options
This item is not returnable. View return policy

Product Code. 12694725

Brand: Avantor J.T.Baker 207806

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 56-85-9
Molecular Formula C5H10N2O3
Packaging Poly Bottle
MDL Number MFCD00008044
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
SMILES N[C@@H](CCC(N)=O)C(O)=O
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
Molecular Weight (g/mol) 146.15
Quantity 1 kg
PubChem CID 5961
ChEBI CHEBI:18050
Formula Weight 146.15
Grade FCC/USP
Chemical Name or Material L-Glutamine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.