missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine tert-butyl ester hydrochloride, 98%

Product Code. p-7055322
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15498556 250 mg 250mg
15440809 1 g 1g
15450809 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498556 Supplier Thermo Scientific Alfa Aesar Supplier No. H63868.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 39741-62-3 | C9H19ClN2O3 | 238.71 g/mol

Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of rhenium-glutamine conjugates. It is mainly used in food and pharma industry.

Solubility
Soluble 0.3gram in 2ml water clear solution.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, acids, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39741-62-3
Molecular Formula C9H19ClN2O3
Molecular Weight (g/mol) 238.71
MDL Number MFCD00039081
InChI Key ZEPNUNKDKQACNC-UHFFFAOYNA-N
Synonym h-gln-otbu.hcl, l-glutamine t-butyl ester hydrochloride, h-gln-otbu hcl, s-tert-butyl 2,5-diamino-5-oxopentanoate hydrochloride, h-gln-otbu hydrochloride, l-glutamine tert-butyl ester hydrochloride, hggln-otbu hcl, gln-otbu.hcl, h-gln-otbucl, glutamine t-butyl ester hydrochloride
PubChem CID 15602445
SMILES Cl.CC(C)(C)OC(=O)C(N)CCC(N)=O

Specifications

Color White
Solubility Information Soluble 0.3gram in 2ml water clear solution.
IUPAC Name tert-butyl 2-amino-4-carbamoylbutanoate hydrochloride
Quantity 250 mg
Formula Weight 238.71
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Glutamine tert-butyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.