missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamine, Cell Culture Reagent

Product Code. 15443319
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Unit Size:
100g
1kg
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443319 100 g 100g
15433319 25 g 25g
15453319 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15443319 Supplier Thermo Scientific Chemicals Supplier No. J60573.22
  • ONLINE EXCLUSIVE PRICE
    £64.06
    £54.65 / 100g
    Save £9.41
    14% Off
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identifiants chimiques

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O

Spécification

pH 5.0 to 6.0
Color White
Beilstein 1723797
Merck Index 14,4471
Quantity 100 g
Formula Weight 146.14
Grade Cell Culture Reagent
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamine

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.