missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, ≥98.5%, Ultrapure

Product Code. p-7082818 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 g
250 g
500 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
100g
1kg
250g
500g
This item is not returnable. View return policy

Product Code. 15875148

Brand: Thermo Scientific J16285.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

CAS: 56-85-9 | C5H10N2O3 | 146.146 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.146
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2 S)-2,5-diamino-5-oxopentanoic acid
SMILES C(CC(=O)N)C(C(=O)O)N

Specifications

Packaging Plastic Bottle
Linear Formula H2NCOCH2CH2CH(NH2)COOH
Beilstein 1723797
Quantity 100 g
Formula Weight 146.14
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.