missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamine-5-13C Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | L-Glutamine-5-13C Hydrochloride |
| Molecular Formula | C413CH11ClN2O3 |
| Quantity | 25 mg |
| Synonym | (S)-2,5-Diamino-5-oxopentanoic Acid-13C Hydrochloride, 2-Aminoglutaramic Acid-13C Hydrochloride, Acustasin-13C Hydrochloride, Aesgen 14-13C Hydrochloride, Cebrogen-13C Hydrochloride, Glumin-13C Hydrochloride, L-2-Aminoglutaramidic Acid-13C Hydrochloride, L-Glutamic Acid γ-αmide-13C Hydrochloride, Levoglutamide-13C Hydrochloride, NSC 27421-13C Hydrochloride, Saforis-13C Hydrochloride, Stimulina-13C Hydrochloride, γ-Glutamine-13C Hydrochloride |
| InChI Formula | InChI=1S/C5H10N2O3.ClH/c6-3(5(9)10)1-2-4(7)8;/h3H,1-2,6H2,(H2,7,8)(H,9,10);1H/t3-;/m0./s1/i4+1; |
| SMILES | N[13C](CC[C@H](N)C(O)=O)=O.Cl |
| Molecular Weight (g/mol) | 183.6 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?