missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, TRC

Product Code. 30004451
Change view
Click to view available options
Quantity:
10 mg
1 g
10 g
Unit Size:
10g
10mg
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004451 10 mg 10mg
30006273 10 g 10g
30004017 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004451 Supplier Toronto Research Chemicals Supplier No. TRCG59696010MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5 H9 N O4
Molecular Weight (g/mol) 147.13
Synonym Glutamic acid, L-Glutamic acid, Glutamic acid, L- (7CI,8CI), (+)-L-Glutamic acid, (2S)-2-Aminopentanedioic acid, (S)-(+)-Glutamic acid, (S)-2-Amino-1,5-pentanedioic acid, (S)-2-Aminopentanedioic acid, (S)-Glutamic acid, 1-Aminopropane-1,3-dicarboxylic acid, 2-Aminoglutaric acid, 2-Aminopentanedioic acid, Aciglut, E 620, Glusate, Glutacid, Glutamic acid, Glutamicol, Glutamidex, Glutaminic acid, Glutaminol, Glutaton, L-(+)-Glutamic acid, L-Glutaminic acid, L-α-Aminoglutaric acid, NSC 143503, Pentanedioic acid, 2-amino-, (S)-, Sortem, l-Glutaminic acid, α-Aminoglutaric acid, α-Glutamic acid
IUPAC Name (2S)-2-aminopentanedioic acid
SMILES N[C@@H](CCC(=O)O)C(=O)O

Specifications

InChI Formula InChI=1S/C5H9NO4/c6-3(5(9)10)1-2-4(7)8/h3H,1-2,6H2,(H,7,8)(H,9,10)/t3-/m0/s1
Quantity 10 mg
Formula Weight 147.0532
Percent Purity >95%
Shipping Condition Room Temperature
Chemical Name or Material L-Glutamic acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.