missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals L(+)-Glutamic acid monosodium salt monohydrate, 99%
£47.20 - £267.00
Chemical Identifiers
CAS | 6106-04-03 |
---|---|
Molecular Formula | C5H8NNaO4·H2O |
Molecular Weight (g/mol) | 187.13 |
MDL Number | MFCD00150138 |
InChI Key | GJBHGUUFMNITCI-QTNFYWBSSA-M |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10265390
|
Thermo Scientific Chemicals
119940010 |
1 kg | Plastic Bottle |
£47.20
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10142770
|
Thermo Scientific Chemicals
119940025 |
2.5 kg | Plastic Bottle |
£92.50
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10237543
|
Thermo Scientific Chemicals
119940100 |
10 kg | Plastic Drum |
£267.00
10kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6106-04-03 | |
187.13 | |
GJBHGUUFMNITCI-QTNFYWBSSA-M |
C5H8NNaO4·H2O | |
MFCD00150138 |
Specifications
232.0°C | |
White | |
0.99 | |
Authentic | |
MFCD00150138 | |
l-thyroxine sodium salt pentahydrate, l-thyroxine sodium pentahydrate, sodium l-thyroxine pentahydrate, sodium levothyroxine, levothyroxine sodium, 3-4-4-hydroxy-3,5-diiodophenoxy-3,5-diiodophenyl-l-alanine sodium salt, levothyroxine sodium pentahydrate, l-tyrosine, o-4-hydroxy-3,5-diiodophenyl-3,5-diiodo-, sodium salt, pentahydrate, o-4-hydroxy-3,5-diiodophenyl-3,5-diiodo-l-tyrosine sodium salt pentahydrate | |
GJBHGUUFMNITCI-QTNFYWBSSA-M | |
sodium (4S)-4-amino-4-carboxybutanoate hydrate | |
187.13 | |
500ppm max | |
99% | |
Crystalline Powder or Crystals |
6106-04-03 | |
2000ppm max | |
C5H8NNaO4·H2O | |
NaO2CCH2CH2CH(NH2)CO2H·H2O | |
0.5% max. (100°C, 5 hrs) | |
Solubility in water: soluble. Other solubilities: sparingly soluble in alcohol | |
O.[Na+].N[C@@H](CCC([O-])=O)C(O)=O | |
+ 25.00 | |
23665037 | |
187.13 | |
+ 25.00 (20.00°C c=10,2 N HCl) | |
L(+)-Glutamic acid monosodium salt monohydrate |
RUO – Research Use Only