missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid monosodium salt monohydrate, 98+%

Product Code. 11409743
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11409743 1000 g 1000g
11419743 250 g 250g
11429743 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11409743 Supplier Thermo Scientific Alfa Aesar Supplier No. A12919.0B
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 6106-04-3 | C5H10NNaO5 | 187.13 g/mol

L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid monosodium salt monohydrate is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals. Also used to intensify flavors of foods.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6106-04-3
Molecular Formula C5H10NNaO5
Molecular Weight (g/mol) 187.13
MDL Number MFCD00150138
InChI Key GJBHGUUFMNITCI-QTNFYWBSSA-M
Synonym monosodium glutamate, l-glutamic acid monosodium salt monohydrate, p.a, l-glutamic acid monosodium salt monohydrate nt, l-glutamic acid monosodium salt monohydrate, saj special grade, monosodium glutamate, united states pharmacopeia usp reference standard, l-glutamic acid, monosodium salt, low endotoxin, pharmagrade, manufactured under appropriate controls for use as a raw material in pharma or biopharmaceutical production., suitable for cell culture
PubChem CID 87090819
IUPAC Name (2S)-2-aminopentanedioic acid;sodium;hydrate
SMILES C(CC(=O)O)C(C(=O)O)N.O.[Na]

Specifications

Melting Point 228°C (decomposition)
Beilstein 4164348
Solubility Information Soluble in water.
Quantity 1000 g
Formula Weight 187.14 (169.12 Anhydrous)
Optical Rotation +22° (c=1 in 6N HCl)
Percent Purity ≥98%
Chemical Name or Material L-Glutamic acid monosodium salt monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.