missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid Monosodium Salt Monohydrate, 98+%, Pure, Fisher Chemical

Product Code. 10665682 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 g
Packaging:
Plastic powder jar
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10665682 250 g Plastic powder jar 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10665682 Supplier Fisher Chemical Supplier No. G/0513/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Melting Point 165°C
CAS 142-47-2
Color White
Physical Form Solid
Quantity 250 g
Molecular Formula C5H8NNaO4
MDL Number MFCD00150138
Synonym natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt
InChI Key LPUQAYUQRXPFSQ-UHFFFAOYNA-M
SMILES [Na+].NC(CCC(O)=O)C([O-])=O
IUPAC Name (2S)-2-aminopentanedioic acid;sodium
Molecular Weight (g/mol) 169.11
PubChem CID 86748263
Formula Weight 169.111g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.