missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid monosodium salt, MedChemExpress

Product Code. 16411746
Encompass_Preferred
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 16411746

Brand: Medchem Express HY14608A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress L-Glutamic acid monosodium salt acts as an excitatory transmitter and an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA). (S)-Glutamic acid shows a direct activating effect on the release of DA from dopaminergic terminals.

L-Glutamic acid monosodium salt acts as an excitatory transmitter and an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA). (S)-Glutamic acid shows a direct activating effect on the release of DA from dopaminergic terminals.

Specifications

Chemical Name or Material L-Glutamic acid monosodium salt
Physical Form Crystals
CAS 142-47-2
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C5H8NNaO4
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information H2O : 7.14 mg/mL (42.22 mM; Need ultrasonic)
SMILES O=C(O[Na])CC[C@H](N)C(O)=O
Molecular Weight (g/mol) 169.11
Formula Weight 169.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.