missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid monosodium salt, MedChemExpress

Product Code. 16466506
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16466506 100 mg 100mg
16411746 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16466506 Supplier Medchem Express Supplier No. HY14608A
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-Glutamic acid monosodium salt acts as an excitatory transmitter and an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA). (S)-Glutamic acid shows a direct activating effect on the release of DA from dopaminergic terminals.

L-Glutamic acid monosodium salt acts as an excitatory transmitter and an agonist at all subtypes of glutamate receptors (metabotropic, kainate, NMDA, and AMPA). (S)-Glutamic acid shows a direct activating effect on the release of DA from dopaminergic terminals.

Specifications

Chemical Name or Material L-Glutamic acid monosodium salt
Physical Form Crystals
CAS 142-47-2
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H8NNaO4
For Use With (Application) Neuroscience-Neurodegeneration
Solubility Information H2O : 7.14 mg/mL (42.22 mM; Need ultrasonic)
SMILES O=C(O[Na])CC[C@H](N)C(O)=O
Molecular Weight (g/mol) 169.11
Formula Weight 169.11
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.