missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamic acid hydrochloride, 99%

Product Code. 10091860 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic Bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091860 500 g Glass Bottle 500g
10286210 100 g Glass Bottle 100g
10137943 2.5 kg Plastic Bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091860 Supplier Thermo Scientific Chemicals Supplier No. 165235000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138-15-8 | C5H10ClNO4 | 183.588 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-15-8
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00012619
InChI Key RPAJSBKBKSSMLJ-DFWYDOINSA-N
Synonym l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin
PubChem CID 2723891
IUPAC Name (2S)-2-aminopentanedioic acid;hydrochloride
SMILES C(CC(=O)O)C(C(=O)O)N.Cl

Specifications

Ammonium 200ppm max.
Arsenic (As) 1ppm max.
pH 1 to 2.5 (2.5% soln. at 25°C)
Color White
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Assay Percent Range 99%
Loss on Drying 0.5% max. (105°C, 5 hrs)
Other Amino Acids 0.5% max. (TLC)
Packaging Glass Bottle
Linear Formula HO2CCH2CH2CH(NH2)CO2H·HCl
Beilstein 04, 488
Specific Rotation +25.2° to +25.8° (20°C, 589nm) (c=10, 2N HCl)
Sulfate 200ppm max.
Merck Index 15, 4505
Transmittance 95% min. (c=10, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Specific Rotation Condition +25.50° (20°C c=10,2N HCl)
Quantity 500 g
Formula Weight 183.59
Percent Purity 98.5 to 101.5%
Physical Form Fine Crystalline Powder or Crystals
Chemical Name or Material L(+)-Glutamic acid hydrochloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • L(+)-Glutamic acid hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.