missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamic acid hydrochloride, 99%

Product Code. 11414154
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11414154

Brand: Thermo Scientific Chemicals A12505.0B

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 138-15-8 | C5H10ClNO4 | 183.588 g/mol

L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

Solubility
Soluble in water (490 g/L).

Notes
Hygroscopic. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138-15-8
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00012619
InChI Key RPAJSBKBKSSMLJ-DFWYDOINSA-N
Synonym l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin
PubChem CID 2723891
IUPAC Name (2S)-2-aminopentanedioic acid;hydrochloride
SMILES C(CC(=O)O)C(C(=O)O)N.Cl

Specifications

Melting Point 203°C (decomposition)
Beilstein 3565569
Merck Index 14,4469
Solubility Information Soluble in water (490g/L).
Density 1.525
Quantity 1000 g
Formula Weight 183.59
Optical Rotation +23° (c=6 in Water)
Sensitivity Hygroscopic
Percent Purity 99%
Chemical Name or Material L-Glutamic acid hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • L-Glutamic acid hydrochloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : L-Glutamic acid, hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.