missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, free acid, ≥99%, MP Biomedicals™

Product Code. 11394239 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11394239 100 g 100g
11304249 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11394239 Supplier MP Biomedicals Supplier No. 0210180080
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 142-47-2
Molecular Formula C5H8NNaO4
Molecular Weight (g/mol) 169.11
MDL Number MFCD00150138
InChI Key LPUQAYUQRXPFSQ-UHFFFAOYNA-M
Synonym natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt
PubChem CID 86748263
IUPAC Name sodium 2-amino-4-carboxybutanoate
SMILES [Na+].NC(CCC(O)=O)C([O-])=O

Specifications

Color White
Assay Percent Range ≥99%
Density 1.538g/mL at 20°C
Quantity 100 g
Formula Weight 187.13
Percent Purity ≥99%
Grade Biochemical
Physical Form Powder
Chemical Name or Material L-Glutamic acid Monosodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.