missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid dimethyl ester hydrochloride, 98+%

Product Code. p-7043970
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11346646

Brand: Thermo Scientific Alfa Aesar L03764.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 23150-65-4 | C7H13ClNO4 | 210.63 g/mol

L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dimethyl ester hydrochloride is used as a pharmaceutical intermediate.

Solubility
Soluble in water, methanol (50 mg/ml).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23150-65-4
Molecular Formula C7H13ClNO4
Molecular Weight (g/mol) 210.63
MDL Number MFCD00038879
InChI Key BPHCSYSXOKTCMA-UHFFFAOYNA-N
Synonym l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride
PubChem CID 12917568
IUPAC Name dimethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES [Cl].COC(=O)CCC(N)C(=O)OC

Specifications

Melting Point 99°C to 103°C
Beilstein 4238829
Solubility Information Soluble in water,methanol (50mg/ml).
Quantity 5 g
Odor Odorless
Formula Weight 211.65
Optical Rotation +26° (c=5 in Water)
Sensitivity Hygroscopic
Percent Purity ≥98%
Chemical Name or Material L-Glutamic acid dimethyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.