missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid diethyl ester hydrochloride, 98%

Product Code. 15465908
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Product Code. Quantity unitSize
15465908 25 g 25g
15455908 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15465908

Brand: Thermo Scientific Alfa Aesar H59551.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1118-89-4 | C9H18ClNO4 | 239.696 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1118-89-4
Molecular Formula C9H18ClNO4
Molecular Weight (g/mol) 239.696
MDL Number MFCD00012509
InChI Key WSEQLMQNPBNMSL-FJXQXJEOSA-N
Synonym l-glutamic acid diethyl ester hydrochloride, diethyl l-glutamate hydrochloride, h-glu oet-oet.hcl, diethyl glutamate hydrochloride, h-glu oet-oet hcl, s-diethyl 2-aminopentanedioate hydrochloride, diethyl l-glutaminate hydrochloride, 1,5-diethyl 2s-2-aminopentanedioate hydrochloride, l-glutamic acid, diethyl ester, hydrochloride, diethyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 73960
IUPAC Name diethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CCOC(=O)CCC(C(=O)OCC)N.Cl

Specifications

Melting Point 108°C to 110°C
Linear Formula C2H5OCOCH2CH2CH(NH2)COOC2H5·HCl
Beilstein 214-270-
Quantity 25 g
Formula Weight 239.7
Optical Rotation +22° (c=1 in H2O)
Percent Purity 98%
Chemical Name or Material L-Glutamic acid diethyl ester hydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.