missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid dibenzyl ester 4-toluenesulfonate, TRC

Product Code. 30240962
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30240962 1 g 1g
30240957 500 mg 500mg
30240780 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30240962 Supplier Toronto Research Chemicals Supplier No. TRCG5970931G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2791-84-6
Molecular Formula C26H29NO7S
Molecular Weight (g/mol) 499.58
Synonym L-Glutamic acid, 1,5-bis(phenylmethyl) ester, 4-methylbenzenesulfonate (1:1) (ACI), L-Glutamic acid, bis(phenylmethyl) ester, 4-methylbenzenesulfonate (9CI), Glutamic acid, dibenzyl ester, p-toluenesulfonate (6CI, 7CI), Glutamic acid, dibenzyl ester, p-toluenesulfonate, L- (8CI), L-Glutamic acid dibenzyl ester p-toluenesulfonate, L-Glutamic acid dibenzyl ester tosylate, Dibenzyl L-glutamate p-toluenesulfonate, Dibenzyl L-glutamate tosylate, H-L-Glu(Obzl)-Obzl tosylate salt, L-glutamic acid dibenzyl ester 4-toluenesulfonate
IUPAC Name dibenzyl (2S)-2-aminopentanedioate;4-methylbenzenesulfonic acid
SMILES Cc1ccc(cc1)S(=O)(=O)O.N[C@@H](CCC(=O)OCc2ccccc2)C(=O)OCc3ccccc3

Specifications

InChI Formula InChI=1S/C19H21NO4.C7H8O3S/c20-17(19(22)24-14-16-9-5-2-6-10-16)11-12-18(21)23-13-15-7-3-1-4-8-15;1-6-2-4-7(5-3-6)11(8,9)10/h1-10,17H,11-14,20H2;2-5H,1H3,(H,8,9,10)/t17-;/m0./s1
Quantity 1 g
Formula Weight 499.1665
Shipping Condition Room Temperature
Chemical Name or Material L-Glutamic acid dibenzyl ester 4-toluenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.