missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid di-tert-butyl ester hydrochloride, 98%

Product Code. 15468456
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15468456

Brand: Thermo Scientific Alfa Aesar H63708.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32677-01-3 | C13H26ClNO4 | 295.804 g/mol

L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid di-tert-butyl ester may be useful in the synthesis of substance P antagonistic peptides.

Solubility
Soluble in methanol (50gmg/ml).

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32677-01-3
Molecular Formula C13H26ClNO4
Molecular Weight (g/mol) 295.804
MDL Number MFCD00058003
InChI Key LFEYMWCCUAOUKZ-FVGYRXGTSA-N
Synonym l-glutamic acid di-tert-butyl ester hydrochloride, h-glu otbu-otbu.hcl, s-di-tert-butyl 2-aminopentanedioate hydrochloride, h-glu otbu-otbu hcl, l-glutamic acid, bis 1,1-dimethylethyl ester, hydrochloride, h-glu otbu-otbu inverted exclamation mark currencyhcl, glutamic acid di-t-butyl ester hydrochloride, s-di-tert-butyl 2-aminopentanedioate hcl, ditert-butyl 2s-2-aminopentanedioate hydrochloride, 1,5-di-tert-butyl 2s-2-aminopentanedioate hydrochloride
PubChem CID 208636
IUPAC Name ditert-butyl (2S)-2-aminopentanedioate;hydrochloride
SMILES CC(C)(C)OC(=O)CCC(C(=O)OC(C)(C)C)N.Cl

Specifications

Beilstein 3573738
Solubility Information Soluble in methanol (50gmg/ml).
Quantity 1 g
Formula Weight 295.8
Percent Purity 98%
Physical Form Solid
Chemical Name or Material L-Glutamic acid di-tert-butyl ester hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.