missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid alpha-(7-amido-4-methylcoumarin)

Product Code. 15446609
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446609 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446609 Supplier Thermo Scientific Alfa Aesar Supplier No. J64847.MF
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 98516-76-8 | C15H16N2O5 | 304.302 g/mol

A fluorogenic substrate for aminopeptidase a, producing a blue fluorescent solution upon cleavage.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

A fluorogenic substrate for aminopeptidase a, producing a blue fluorescent solution upon cleavage.
TRUSTED_SUSTAINABILITY

Specifications

CAS 98516-76-8
Color White
Molecular Formula C15H16N2O5
MDL Number MFCD00051217
Synonym l-glutamic acid alpha-7-amido-4-methylcoumarin, glu-nh-med, 4s-4-amino-5-4-methyl-2-oxochromen-7-yl amino-5-oxopentanoic acid, s-4-amino-5-4-methyl-2-oxo-2h-chromen-7-yl amino-5-oxopentanoic acid, 4s-4-amino-4-4-methyl-2-oxochromen-7-yl carbamoyl butanoic acid, ambotzhaa7670, 7-alpha-glutamyl-4-methylcoumarylamide, h-glu-amc, alpha-glutamyl-7-amino-4-methylcoumarylamide, n-4-methyl-2-oxo-2h-chromen-7-yl-l-, a-glutamine
InChI Key USALUSONPMPEKY-NSHDSACASA-N
SMILES CC1=CC(=O)OC2=C1C=CC(=C2)NC(=O)C(CCC(=O)O)N
IUPAC Name (4S)-4-amino-5-[(4-methyl-2-oxochromen-7-yl)amino]-5-oxopentanoic acid
Molecular Weight (g/mol) 304.302
Quantity 50 mg
PubChem CID 126959
Formula Weight 304.3
Physical Form Powder
Chemical Name or Material L-Glutamic acid alpha-(7-amido-4-methylcoumarin)
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.