missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid a-amide, TRC

Product Code. 30400980
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400980 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30400980 Supplier Toronto Research Chemicals Supplier No. TRCG650140500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Glutamic acid alpha-amide
CAS 636-65-7
Molecular Formula C5H10N2O3
Quantity 500 mg
Synonym L-isoglutamine, 4-amino-L-glutaramic acid, (4S)-4,5-Diamino-4-oxo-pentanoic acid
InChI Formula InChI=1S/C5H10N2O3/c6-3(5(7)10)1-2-4(8)9/h3H,1-2,6H2,(H2,7,10)(H,8,9)/t3-/m0/s1
SMILES N[C@@H](CCC(=O)O)C(=O)N
IUPAC Name (4S)-4,5-diamino-5-oxopentanoic acid
Molecular Weight (g/mol) 146.14
Formula Weight 146.0691
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.