missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, 99+%, Monosodium Salt, MP Biomedicals

Product Code. 11455192 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
Product Code. Quantity unitSize
11455192 100 g 100g
11465192 500 g 500g
11475192 1 kg 1kg
3 options
This item is not returnable. View return policy

Product Code. 11455192

Brand: MP Biomedicals 0219467780

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 142-47-2
Molecular Formula C5H8NNaO4
Molecular Weight (g/mol) 169.11
MDL Number MFCD00150138
InChI Key LPUQAYUQRXPFSQ-UHFFFAOYNA-M
Synonym natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt
PubChem CID 86748263
IUPAC Name sodium 2-amino-4-carboxybutanoate
SMILES [Na+].NC(CCC(O)=O)C([O-])=O

Specifications

Quantity 100 g
Formula Weight 169.1
Grade Cell Culture
Chemical Name or Material L-Glutamic Acid, 99+%, Monosodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.