missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-benzyl ester, 99%

Product Code. 11449727
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11449727 25 g 25g
11429727 1 g 1g
11439727 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11449727 Supplier Thermo Scientific Alfa Aesar Supplier No. B22184.14
 Restricted Item
Estimated Shipment: 24-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 1676-73-9 | C12H15NO4 | 237.255 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1676-73-9
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00002633
InChI Key BGGHCRNCRWQABU-JTQLQIEISA-N
Synonym h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
PubChem CID 122337
IUPAC Name (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N

Specifications

Melting Point 162°C to 165°C
Beilstein 1885646
Quantity 25 g
Formula Weight 237.26
Optical Rotation +23° (c=1 in Acetic Acid)
Percent Purity 99%
Chemical Name or Material L-Glutamic acid 5-benzyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.