missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 1-tert-butyl ester, 97%

Product Code. 15442529
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15442529

Brand: Thermo Scientific Alfa Aesar H65342.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 45120-30-7 | C9H17NO4 | 203.238 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 45120-30-7
Molecular Formula C9H17NO4
Molecular Weight (g/mol) 203.238
MDL Number MFCD00038562
InChI Key QVAQMUAKTNUNLN-LURJTMIESA-N
Synonym h-glu-otbu, s-4-amino-5-tert-butoxy-5-oxopentanoic acid, 1-tert-butyl l-glutamate, l-glutamic acid 1-tert-butyl ester, 4s-4-amino-5-tert-butoxy-5-oxopentanoic acid, glu-otbu, h-gluotbu, ambotzhaa5210, pubchem17225, glutamic acid t-butyl ester
PubChem CID 117505
IUPAC Name (4S)-4-amino-5-[(2-methylpropan-2-yl)oxy]-5-oxopentanoic acid
SMILES CC(C)(C)OC(=O)C(CCC(=O)O)N

Specifications

Melting Point 139°C to 145°C
Color White
Beilstein 2414793
Quantity 5 g
Formula Weight 203.24
Percent Purity 97%
Physical Form Powder
Chemical Name or Material L-Glutamic acid 1-tert-butyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.