missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid 1-Methyl Ester, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | L-Glutamic Acid 1-Methyl Ester |
| CAS | 6384-08-3 |
| Molecular Formula | C6H11NO4 |
| Quantity | 100 mg |
| Synonym | Glutamic Acid Methyl Ester, Glutamic Acid 1-Methyl Ester, Glutamic Acid α-Methyl Ester, L-Glutamic Acid Methyl Ester, L-Glutamic Acid α-Methyl Ester, Methyl (S)-2-Amino-4-carboxybutanoate, Methyl L-Glutamate, α-Methyl L-Glutamate |
| InChI Formula | InChI=1S/C6H11NO4/c1-11-6(10)4(7)2-3-5(8)9/h4H,2-3,7H2,1H3,(H,8,9)/t4-/m0/s1 |
| SMILES | N[C@H](C(OC)=O)CCC(O)=O |
| IUPAC Name | (S)-4-amino-5-methoxy-5-oxopentanoic acid |
| Molecular Weight (g/mol) | 161.16 |
| Formula Weight | 161.07 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction