missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(-)-Glucose, 98%

Product Code. p-3778706 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
25g
5g
This item is not returnable. View return policy

Product Code. 10552061

Brand: Thermo Scientific Chemicals 241922500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 921-60-8 | C6H12O6 | 180.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 921-60-8
Molecular Formula C6H12O6
Molecular Weight (g/mol) 180.16
MDL Number MFCD00148913
InChI Key GZCGUPFRVQAUEE-VANKVMQKSA-N
Synonym l-glucose, l---glucose, levoglucose, l--glucose, 2s,3r,4s,5s-2,3,4,5,6-pentahydroxyhexanal, aldehydo-l-glucose, aldehydo-l-gluco-hexose, d-glucose-13c6,d7, levoglucose usan, glucose, l-isomer
PubChem CID 10954115
ChEBI CHEBI:37626
IUPAC Name (2S,3R,4S,5S)-2,3,4,5,6-pentahydroxyhexanal
SMILES C(C(C(C(C(C=O)O)O)O)O)O

Specifications

Melting Point 153.0°C to 156.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (ELSD) (HPLC)
Packaging Glass bottle
Quantity 250 mg
Specific Rotation Condition − 52.00 (20.00°C c=10,H2O,NH3)
Specific Rotation − 52.00
Solubility Information (10% in water) Clear colorless
Formula Weight 180.16
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material L(-)-Glucose
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.