missing translation for 'onlineSavingsMsg'
Learn More

L-Fuculose (1M in Water), TRC

Product Code. 30394972
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30394972 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30394972 Supplier Toronto Research Chemicals Supplier No. TRCF83613510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 13074-08-3
Molecular Formula C6H12O5
Quantity 10 mg
Synonym 6-Deoxy-L-tagatose
InChI Formula InChI=1S/C6H12O5/c1-3(8)5(10)6(11)4(9)2-7/h3,5-8,10-11H,2H2,1H3/t3-,5+,6-/m0/s1
SMILES C[C@H](O)[C@@H](O)[C@@H](O)C(=O)CO
IUPAC Name (3R,4R,5S)-1,3,4,5-tetrahydroxyhexan-2-one
Molecular Weight (g/mol) 164.16
Formula Weight 164.0685
Concentration or Composition (by Analyte or Components) 1 M
Chemical Name or Material L-Fuculose
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.