missing translation for 'onlineSavingsMsg'
Learn More

L-Formiminoglutamic Acid, TRC

Product Code. 30395566
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30395566 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30395566 Supplier Toronto Research Chemicals Supplier No. TRCF7355581MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Formiminoglutamic Acid
CAS 816-90-0
Purity Grade Notes HPLC
Molecular Formula C6 H10 N2 O
Quantity 1 mg
Synonym N-(Iminomethyl)-L-glutamic Acid, Formimino-L-glutamic Acid, Formiminoglutamic Acid, N-Formimino-L-glutamic Acid, NSC 334078
InChI Formula InChI=1S/C6H10N2O4/c7-3-8-4(6(11)12)1-2-5(9)10/h3-4H,1-2H2,(H2,7,8)(H,9,10)(H,11,12)/t4-/m0/s1
SMILES N\C=N\[C@@H](CCC(=O)O)C(=O)O
IUPAC Name (2S)-2-[(E)-aminomethyleneamino]pentanedioic acid
Molecular Weight (g/mol) 174.16
Formula Weight 174.064
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.