Learn More
Invitrogen™ L-Ficolin Monoclonal Antibody (GN5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA533345
Description
Product is supplied at 1 mL (100 μL/mL). Application Notes: IHC-F - Tissue sections were fixed in acetone and pretreated with peroxidase block for 5 min. in the dark at RT to quench endogenous peroxidases. Western blot - A non-reduced sample treatment and SDS-Page was used. The band size is 35kDa.
L-Ficolin (Ficolin-2) is an innate immunity pattern recognition molecule found in human serum. L-Ficolin binds to distinct pathogen-associated molecular patterns (PAMP), such as carbohydrates (GlcNAc), lipoteichoic acid and acetylated groups thereby facilitating phagocytosis and activation of complement through the lectin route using the same serine proteases as mannose-binding lectin (MBL) named MASP ((MBL)-associated serine protease).
Specifications
| L-Ficolin | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| Q15485 | |
| FCN2 | |
| Protein G | |
| RUO | |
| 2220 | |
| 4°C | |
| Liquid |
| ELISA, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot | |
| GN5 | |
| Unconjugated | |
| FCN2 | |
| 37 kDa elastin-binding protein; Collagen/fibrinogen domain-containing protein 2; EBP-37; FCN2; Fcnb; FCNL; ficolin (collagen/fibrinogen domain containing lectin) 2 (hucolin); ficolin 2; ficolin B; ficolin-2; Ficolin-B; ficolin-beta; Hucolin; L-ficolin; P35; RP11-263F14.2; Serum lectin p35 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.