missing translation for 'onlineSavingsMsg'
Learn More

L-(-)-Epinephrine, TRC

Product Code. 30041067
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30041067 100 mg 100mg
30008965 1 g 1g
30005700 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041067 Supplier Toronto Research Chemicals Supplier No. TRCE588580100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51-43-4
Formula Weight 183.0895
InChI Formula InChI=1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/t9-/m0/s1
IUPAC Name 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
Molecular Formula C9 H13 N O3
Molecular Weight (g/mol) 183.2
Percent Purity >95%
SMILES CNC[C@H](O)c1ccc(O)c(O)c1
Synonym Noradrenaline Tartrate Imp. A (EP), 1,2-Benzenediol, 4-[(1R)-1-hydroxy-2-(methylamino)ethyl]-, 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, (R)-, Benzyl alcohol, 3,4-dihydroxy-α-[(methylamino)methyl]-, (-)- (8CI), 4-[(1R)-1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol, (-)-(R)-Epinephrine, (-)-3,4-Dihydroxy-α-[2-(methylamino)ethyl]benzyl alcohol, (-)-Adrenaline, (-)-Epinephrine, (R)-Adrenaline, (R)-Epinephrine, Adnephrine, Adrenal, Adrenalin, Adrenaline, Adrenine, Adrin, Ana-Kit, Bosmin, Bronkaid Mist, Chelafrin, Epifrin, Epiglaufrin, Epinefrina, Epinephran, Epinephrine, Epipen, Epirenan, Eppy, Exadrin, Glauposine, Hemisine, Hemostasin, Hemostatin, Hypernephrin, Isoptoepinal, L-Adrenaline, L-Methylaminoethanolcatechol, Levoepinephrine, Levorenen, Levorenin, Levorenine, Levoreninum, Lyodrin, Methylarterenol, Mucidrina, N-[(2R)-2-(3,4-Dihydroxyphenyl)-2-hydroxyethyl]-N-methylamine, NSC 62786, Nephridine, Nieraline, Paranephrin, Primatene Mist, R-(-)-Epinephrine, Renaglandin, Renaleptine, Renalina, Renoform, Renostypticin, Renostyptin, Scurenaline, Simplene, Styptirenal, Supracapsulin, Supranephrane, Suprarenaline, Suprarenin, Surrenine, Sus-phrine, Takamina, Vasoconstrictine, Vasotonin, l-1-(3,4-Dihydroxyphenyl)-2-methylaminoethanol, l-Adrenaline, l-Epinephrine, l-Epirenamine
Chemical Name or Material L-Adrenaline
Grade HPLC
Show More Show Less

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.