missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-DOPA Methyl Ester Hydrochloride, TRC
Specifications
Specifications
| Chemical Name or Material | L-DOPA Methyl Ester Hydrochloride |
| CAS | 1421-65-4 |
| Molecular Formula | C10 H13 N O4 . Cl H |
| Quantity | 5 g |
| Synonym | L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (1:1), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride (6CI,7CI), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride, L- (8CI), L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (9CI), 3,4-Dihydroxyphenyl-L-alanine methyl ester hydrochloride, L-DOPA Me ester hydrochloride, L-Dopa methyl ester hydrochloride, Levodopa methyl ester hydrochloride, Methyl L-3-(3,4-dihydroxyphenyl)alaninate hydrochloride, ST 41769 |
| InChI Formula | InChI=1S/C10H13NO4.ClH/c1-15-10(14)7(11)4-6-2-3-8(12)9(13)5-6;/h2-3,5,7,12-13H,4,11H2,1H3;1H/t7-;/m0./s1 |
| SMILES | Cl.COC(=O)[C@@H](N)Cc1ccc(O)c(O)c1 |
| IUPAC Name | methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride |
| Molecular Weight (g/mol) | 247.68 |
| Formula Weight | 247.0611 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?