missing translation for 'onlineSavingsMsg'
Learn More

L-DOPA Methyl Ester Hydrochloride, TRC

Product Code. 30383666
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383666 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30383666 Supplier Toronto Research Chemicals Supplier No. TRCD533770500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-DOPA Methyl Ester Hydrochloride
CAS 1421-65-4
Molecular Formula C10 H13 N O4 . Cl H
Quantity 500 mg
Synonym L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (1:1), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride (6CI,7CI), Alanine, 3-(3,4-dihydroxyphenyl)-, methyl ester, hydrochloride, L- (8CI), L-Tyrosine, 3-hydroxy-, methyl ester, hydrochloride (9CI), 3,4-Dihydroxyphenyl-L-alanine methyl ester hydrochloride, L-DOPA Me ester hydrochloride, L-Dopa methyl ester hydrochloride, Levodopa methyl ester hydrochloride, Methyl L-3-(3,4-dihydroxyphenyl)alaninate hydrochloride, ST 41769
InChI Formula InChI=1S/C10H13NO4.ClH/c1-15-10(14)7(11)4-6-2-3-8(12)9(13)5-6;/h2-3,5,7,12-13H,4,11H2,1H3;1H/t7-;/m0./s1
SMILES Cl.COC(=O)[C@@H](N)Cc1ccc(O)c(O)c1
IUPAC Name methyl (2S)-2-amino-3-(3,4-dihydroxyphenyl)propanoate;hydrochloride
Molecular Weight (g/mol) 247.68
Formula Weight 247.0611
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.