missing translation for 'onlineSavingsMsg'
Learn More

L-DOPA, MedChemExpress

Product Code. 16432519
Encompass_Preferred
Change view
Click to view available options
Quantity:
200 mg
1 g
Unit Size:
1g
200mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432519 1 g 1g
16421506 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16432519 Supplier Medchem Express Supplier No. HYN0304
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress L-DOPA (Levodopa) is an orally active metabolic precursor of neurotransmitters dopamine. L-DOPA can cross the blood-brain barrier and is converted into dopamine in the brain. L-DOPA has anti-allodynic effects and the potential for Parkinson's disease.

L-DOPA (Levodopa) is an orally active metabolic precursor of neurotransmitters dopamine. L-DOPA can cross the blood-brain barrier and is converted into dopamine in the brain. L-DOPA has anti-allodynic effects and the potential for Parkinson's disease.

Specifications

Chemical Name or Material L-DOPA
CAS 59-92-7
Color White
Purity Grade Notes Research
Molecular Formula C9H11NO4
Quantity 1 g
Synonym Levodopa 3,4-Dihydroxyphenylalanine
Solubility Information 0.1 M HCL : 20 mg/mL (101.43 mM; ultrasonic and warming and adjust pH to 2 with HCl and heat to 60°C) ∣H2O : 1 mg/mL (5.07 mM; Need ultrasonic)
SMILES N[C@@H](CC1=CC(O)=C(C=C1)O)C(O)=O
Molecular Weight (g/mol) 197.19
Formula Weight 197.19
Percent Purity 99.11%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.