missing translation for 'onlineSavingsMsg'
Learn More

L-Dehydroascorbic acid, 96%

Product Code. 15720849 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15720849 1 g 1g
15730849 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15720849 Supplier Thermo Scientific Acros Supplier No. 250930010
  • ONLINE EXCLUSIVE PRICE
    £107.00
    £96.65 / 1g
    Save £10.35
    9% Off
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 490-83-5 | C6H6O6 | 174.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-83-5
Molecular Formula C6H6O6
Molecular Weight (g/mol) 174.11
InChI Key SBJKKFFYIZUCET-UHFFFAOYNA-N
IUPAC Name 5-(1,2-dihydroxyethyl)oxolane-2,3,4-trione
SMILES OCC(O)C1OC(=O)C(=O)C1=O

Specifications

Melting Point 228.0°C
Assay Percent Range 95.0 % min. (ELSD) (HPLC)
Quantity 1 g
Formula Weight 174.11
Chemical Name or Material L-Dehydroascorbic acid
compliance-icons
Product Identifier
  • L-Dehydroascorbic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
  • MIXTURE LIST-Contain: L-Dehydroascorbic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.