missing translation for 'onlineSavingsMsg'
Learn More

L-Dehydro Ascorbic Acid (>90%), TRC

Product Code. 30133349
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133349 1 g 1g
17938584 500 mg 500mg
17957614 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133349 Supplier Toronto Research Chemicals Supplier No. TRCD2292501G
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 490-83-5
Molecular Formula C6 H6 O6
Molecular Weight (g/mol) 174.11
Synonym L-threo-2,3-Hexodiulosonic acid γ-lactone, Dehydroascorbic acid, L- (8CI), DHAA, Dehydro-L-ascorbic acid, Dehydroascorbic acid, L-Ascorbic acid, dehydro-, L-Ascorbic acid, oxidized, L-Dehydroascorbic acid, L-threo-Dehydroascorbic acid, Oxidized ascorbic acid, Oxidized vitamin C
IUPAC Name (5R)-5-[(1S)-1,2-dihydroxyethyl]oxolane-2,3,4-trione
SMILES OC[C@H](O)[C@H]1OC(=O)C(=O)C1=O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6O6/c7-1-2(8)5-3(9)4(10)6(11)12-5/h2,5,7-8H,1H2/t2-,5+/m0/s1
Formula Weight 174.0164
Percent Purity >95%
Chemical Name or Material Dehydroascorbic Acid
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.