missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine disodium salt, 98+%

Product Code. 15404719
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15404719

Brand: Thermo Scientific Alfa Aesar J62310.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 64704-23-0 | C6H10N2Na2O4S2 | 284.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64704-23-0
Molecular Formula C6H10N2Na2O4S2
Molecular Weight (g/mol) 284.26
MDL Number MFCD00037206
InChI Key PLVPMKWGXOOSKL-UHFFFAOYNA-L
Synonym disodium 2-amino-3-2r-2-amino-2-carboxylatoethyl disulfanyl propanoate
PubChem CID 71607258
IUPAC Name disodium;2-amino-3-[[(2R)-2-amino-2-carboxylatoethyl]disulfanyl]propanoate
SMILES [Na+].[Na+].NC(CSSCC(N)C([O-])=O)C([O-])=O

Specifications

Color Brown
Quantity 250 g
Optical Rotation +170° (c=2 in 1N HCl)
Formula Weight 284.3
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material L-Cystine disodium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.