missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine-dimethyl Ester Dihydrochloride, TRC

Product Code. 30359400
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30359400 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30359400 Supplier Toronto Research Chemicals Supplier No. TRCC9979005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Cystine-dimethyl Ester Dihydrochloride
CAS 32854-09-4
Molecular Formula C8 H16 N2 O4 S2 . 2 Cl H
Quantity 5 g
Synonym L-Cystine, 1,1'-dimethyl ester, hydrochloride (1:2) (ACI), L-Cystine, dimethyl ester, dihydrochloride (9CI), Cystine, dimethyl ester, dihydrochloride, L- (6CI, 7CI, 8CI), L-Cystine dihydrochloride methyl ester, Cystine dimethyl ester dihydrochloride, NSC 161614, L-Cystine dimethyl ester dihydrochloride, Dimethyl L-cystinate dihydrochloride, Methyl (2R)-2-amino-3-[[(2R)-2-amino-3-methoxy-3-oxo-propyl]disulfanyl]propanoate dihydrochloride
InChI Formula InChI=1S/C8H16N2O4S2.2ClH/c1-13-7(11)5(9)3-15-16-4-6(10)8(12)14-2;;/h5-6H,3-4,9-10H2,1-2H3;2*1H/t5-,6-;;/m0../s1
SMILES Cl.Cl.COC(=O)[C@@H](N)CSSC[C@H](N)C(=O)OC
IUPAC Name methyl (2R)-2-amino-3-[[(2R)-2-amino-3-methoxy-3-oxopropyl]disulfanyl]propanoate;dihydrochloride
Molecular Weight (g/mol) 341.28
Formula Weight 340.0085
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.