missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine dimethyl ester dihydrochloride, 98%

Product Code. 15419828
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15419828 1 g 1g
15429828 5 g 5g
15439828 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15419828 Supplier Thermo Scientific Alfa Aesar Supplier No. H63234.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 32854-09-4 | C8H18Cl2N2O4S2 | 341.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32854-09-4
Molecular Formula C8H18Cl2N2O4S2
Molecular Weight (g/mol) 341.26
MDL Number MFCD00012490,MFCD00012490,MFCD09951837,MFCD00012490
InChI Key QKWGUPFPCRKKMQ-UHFFFAOYNA-N
Synonym h-cys-ome 2.2hcl, dimethyl l-cystinate dihydrochloride, h-d-cys-ome 2 2hcl, methyl 2-amino-3-r-2-amino-3-methoxy-3-oxopropyl disulfanyl propanoate dihydrochloride, d-cystine bis methyl ester dihydrochloride, d-cystine dimethyl ester dihydrochloride, h-d-cys-ome 2 hcl, c8h16n2o4s2.2hcl, dimethyl l-cystinate 2hcl, h-d-cys-ome 2 . 2 hcl
PubChem CID 71463656
IUPAC Name methyl (2S)-2-amino-3-[[(2S)-2-amino-3-methoxy-3-oxopropyl]disulfanyl]propanoate;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].COC(=O)C(N)CSSCC(N)C(=O)OC

Specifications

Melting Point 182°C to 183°C (decomposition)
Color White
Quantity 1 g
Beilstein 4727524
Formula Weight 341.28
Percent Purity 98%
Physical Form Powder
Chemical Name or Material L-Cystine dimethyl ester dihydrochloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.