missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine Dihydrochloride, TRC

Product Code. 30366216
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 30366216

Brand: Toronto Research Chemicals TRCC997903250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material L-Cystine Dihydrochloride
CAS 30925-07-6
Molecular Formula C6H12N2O4S2 . 2(HCl)
Quantity 250 mg
Synonym L-Cystine, hydrochloride (1:2) (ACI), L-Cystine, dihydrochloride (9CI), Cystine, dihydrochloride, L- (8CI), Cystine dihydrochloride, L-Cystine Dihydrochloride, (2R)-2-Amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid dihydrochloride, 3,3'-Dithiobis[(2R)-2-aminopropanoic acid] dihydrochloride, (R,R)-Bis(β-amino-β-carboxyethyl) disulfide dihydrochloride
InChI Formula InChI=1S/C6H12N2O4S2.2ClH/c7-3(5(9)10)1-13-14-2-4(8)6(11)12;;/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12);2*1H/t3-,4-;;/m0../s1
SMILES Cl.Cl.N[C@@H](CSSC[C@H](N)C(=O)O)C(=O)O
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride
Molecular Weight (g/mol) 313.22
Formula Weight 311.9772
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.