missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cystine Dihydrochloride, TRC
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
Specifications
Specifications
| Chemical Name or Material | L-Cystine Dihydrochloride |
| CAS | 30925-07-6 |
| Molecular Formula | C6H12N2O4S2 . 2(HCl) |
| Quantity | 500 mg |
| Synonym | L-Cystine, hydrochloride (1:2) (ACI), L-Cystine, dihydrochloride (9CI), Cystine, dihydrochloride, L- (8CI), Cystine dihydrochloride, L-Cystine Dihydrochloride, (2R)-2-Amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid dihydrochloride, 3,3'-Dithiobis[(2R)-2-aminopropanoic acid] dihydrochloride, (R,R)-Bis(β-amino-β-carboxyethyl) disulfide dihydrochloride |
| InChI Formula | InChI=1S/C6H12N2O4S2.2ClH/c7-3(5(9)10)1-13-14-2-4(8)6(11)12;;/h3-4H,1-2,7-8H2,(H,9,10)(H,11,12);2*1H/t3-,4-;;/m0../s1 |
| SMILES | Cl.Cl.N[C@@H](CSSC[C@H](N)C(=O)O)C(=O)O |
| IUPAC Name | (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride |
| Molecular Weight (g/mol) | 313.22 |
| Formula Weight | 311.9772 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction