missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine Dihydrochloride, MP Biomedicals™

Product Code. 11431952 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g

Product Code. 11431952

Brand: MP Biomedicals 0210557680

Please to purchase this item. Need a web account? Register with us today!

Used as a cell culture supplement. L-Cystine Dihydrochloride, MP Biomedicals is a natural proteinogenic amino acid sulfide dimer of L-cysteine. It is also used to characterize and identify specific amino acid transport systems in cells such as system syst

TRUSTED_SUSTAINABILITY

Specifications

CAS 30925-07-6
Melting Point 228°C to 232°C (decomposition)
Molecular Formula C6H14Cl2N2O4S2
Quantity 100 g
Synonym l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis
Solubility Information Soluble in water.
InChI Key HHGZUQPEIHGQST-RGVONZFCSA-N
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride
Molecular Weight (g/mol) 313.208
PubChem CID 21121987
Formula Weight 313.2
Physical Form Powder
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.