missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine dihydrochloride, 98%

Product Code. 15480317
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15480317

Brand: Thermo Scientific Alfa Aesar J63717.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 30925-07-6 | C6H14Cl2N2O4S2 | 313.208 g/mol

Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Non-animal sourced L-cystine is used as a cell culture media component for the commercial biomanufacture of therapeutic recombinant proteins and monoclonal antibodies. L-cystine is used as a cell culture supplement.

Solubility
Freely soluble in Dilute hydrochloric acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30925-07-6
Molecular Formula C6H14Cl2N2O4S2
Molecular Weight (g/mol) 313.208
MDL Number MFCD00070399
InChI Key HHGZUQPEIHGQST-RGVONZFCSA-N
Synonym l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis
PubChem CID 21121987
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl

Specifications

Melting Point 228°C to 232°C
Color White to Yellow
Solubility Information Freely soluble in Dilute hydrochloric acid.
Quantity 250 g
Formula Weight 313.22
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Cystine dihydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.