missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford L-Cystine Dihydrochloride
Click to view available options
Quantity:
5 kg
Unit Size:
5kg
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
- CAS Number: 30925-07-6
- Molecular Formula: C6H12N2O4S2 2HCl
- Molecular Weight: 313.2
Specifications
Specifications
| Chemical Name or Material | L-Cystine, Dihydrochloride |
| CAS | 30925-07-6 |
| Molecular Formula | C6H12N2O4S2 - 2 HCl |
| Quantity | 5 kg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction