missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Cystine dihydrochloride, 100.3%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| CAS | 30925-07-6 |
| Molecular Formula | C6H14Cl2N2O4S2 |
| Synonym | l-cystine dihydrochloride, l---cystine dihydrochloride, cystine, dihydrochloride, h-cys-oh 2.2hcl, c6h12n2o4s2.2hcl, h-cys-oh 2 . 2hcl, l-cystine dihydrochloride tlc, l-cystine dihydrochloride, cell culture reagent, 2r-2-amino-3-2r-2-amino-2-carboxyethyl disulfanyl propanoic acid dihydrochloride, l-cystine dihydrochloride, from non-animal source, bioreagent, suitable for cell culture dry basis |
| InChI Key | HHGZUQPEIHGQST-RGVONZFCSA-N |
| SMILES | C(C(C(=O)O)N)SSCC(C(=O)O)N.Cl.Cl |
| IUPAC Name | (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid;dihydrochloride |
| Molecular Weight (g/mol) | 313.208 |
| Quantity | 5 g |
| PubChem CID | 21121987 |
| Chemical Name or Material | L-Cystine dihydrochloride |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction