missing translation for 'onlineSavingsMsg'
Learn More

L-Cystine, Cell Culture Reagent

Product Code. p-7042195
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
500g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15424209 10 g 10g
15434209 100 g 100g
15444209 500 g 500g
15454209 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15424209 Supplier Thermo Scientific Alfa Aesar Supplier No. J61651.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 56-89-3 | C6H12N2O4S2 | 240.292 g/mol

L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-cysteine acts as a major source of sulfur in cell culture. Cysteine has been used in pulse-chase experiments to study protein expression from an inducible promoter. L-Cysteine is used to supplement cell culture media.

Solubility
It is soluble in water and HCL.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-89-3
Molecular Formula C6H12N2O4S2
Molecular Weight (g/mol) 240.292
MDL Number MFCD00064228
InChI Key LEVWYRKDKASIDU-IMJSIDKUSA-N
Synonym l-cystine, cystine, l-dicysteine, l-cystin, cystine acid, cysteine disulfide, oxidized l-cysteine, beta,beta'-dithiodialanine, h-cys-oh 2, l-cysteine disulfide
PubChem CID 67678
ChEBI CHEBI:16283
IUPAC Name (2R)-2-amino-3-[[(2R)-2-amino-2-carboxyethyl]disulfanyl]propanoic acid
SMILES C(C(C(=O)O)N)SSCC(C(=O)O)N

Specifications

Melting Point >240°C (decomposition)
Color White
Beilstein 1728094
Merck Index 14,2782
Solubility Information It is soluble in water and HCL.
Density 1.68
Quantity 10 g
Formula Weight 240.3
Optical Rotation −219° (c=1 in 1N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Cystine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.